Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, puriss., ≥99.7% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120579 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
160 kg
Packaging:
Metal Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-577 2.5 L Poly Bottle
60-045-580 4 x 2.5 L Poly Bottle
60-045-576 160 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-577 Supplier Honeywell Chemicals Supplier No. 242292.5L
Only null left
Add to Cart
Add to Cart

Puriss., meets analytical specification of Ph Eur,≥99.7% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791 to 0.793g/mL at 20°C, 0.791g/mL at 25°C
Boiling Point 64.7°C
Quantity 2.5 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.7% (GC)
Grade puriss.
Chemical Name or Material Methanol, Meets analytical specification of Ph Eur
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
Precautionary Statement H225-H301 + H311 + H331-H370
DOTInformation Class 3 - PG 2 - Methanol
EINECSNumber 200-659-6
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.