Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methicillin Resistant Staphylococcus Aureus Antibody (332/423), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP24428A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24428A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24428A94 Supplier Novus Biologicals Supplier No. NBP254428AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methicillin Resistant Staphylococcus Aureus Monoclonal specifically detects Methicillin Resistant Staphylococcus Aureus in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Methicillin Resistant Staphylococcus Aureus
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 332/423
Conjugate Alexa Fluor 594
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Methicillin-resistant Staphylococcus aureus (MRSA)
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.