Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methicillin Resistant Staphylococcus Aureus Antibody (332/423), Novus Biologicals™
SDP

Catalog No. NB253147100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253147100 100 μg
NB25314720 20 μg
NB253147200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253147100 Supplier Novus Biologicals Supplier No. NBP253147100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methicillin Resistant Staphylococcus Aureus Monoclonal specifically detects Methicillin Resistant Staphylococcus Aureus in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Methicillin Resistant Staphylococcus Aureus
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 332/423
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Host Species Mouse
Immunogen Methicillin-resistant Staphylococcus aureus (MRSA)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.