Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methionine Aminopeptidase 2/METAP2 Antibody (001), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB040309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040309 Supplier Novus Biologicals Supplier No. NBP290622UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Methionine Aminopeptidase 2/METAP2 Monoclonal specifically detects Methionine Aminopeptidase 2/METAP2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Methionine Aminopeptidase 2/METAP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Methionine Aminopeptidase 2/METAP2 (O08663; Ala2-Tyr478).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10988
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.