Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methionyl tRNA synthetase Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290262 Supplier Novus Biologicals Supplier No. NBP222312APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Methionyl tRNA synthetase Polyclonal antibody specifically detects Methionyl tRNA synthetase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Methionyl tRNA synthetase
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias cytosolic methionyl-tRNA synthetase, EC 6.1.1.10, FLJ35667, methionine tRNA ligase 1, cytoplasmic, Methionine--tRNA ligase, methionyl-tRNA synthetase, methionyl-tRNA synthetase, cytoplasmic, METRS, MRS, MTRNS
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 175 to 225 of human Methionyl-tRNA Synthetase using the numbering given in entry NP_004981.2 (GeneID 4141).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4141
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.