Learn More
Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech.
Description
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.
Solubility
Miscible with acetone, benzene, naphtha and xylene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Density | 1.232 |
| Boiling Point | 137°C to 140°C (10 mmHg) |
| Flash Point | 135°C (275°F) |
| Refractive Index | 1.5052 |
| Quantity | 500 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with acetone,benzene,naphtha and xylene. |
| Formula Weight | 178.19 |
| Grade | Technical |
| Chemical Name or Material | Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers |
Safety and Handling
| Hazard Category | H302-H315-H317-H318-H331-H334 |
| Hazard Statement | GHS H Statement H301-H334-H332-H315-H319-H335 Toxic if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P311-P330-P333+P313-P362-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 246-644-8 |
| RTECSNumber | RB9100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.