Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methoxyacetic acid, 97%

Click to view available options
Quantity:
50g
250g
1000g
625-45-6
C3H6O3
90.078
MFCD00004308
RMIODHQZRUFFFF-UHFFFAOYSA-N
methoxyacetic acid,acetic acid, methoxy,methoxyethanoic acid,acetic acid, 2-methoxy,ch3och2cooh,methoxyacetlc acid,unii-f11t1h7q7w,methoxy acetic acid,methoxy-acetic acid,ccris 6518
12251
2-methoxyacetic acid
COCC(=O)O
625-45-6
C3H6O3
90.078
MFCD00004308
RMIODHQZRUFFFF-UHFFFAOYSA-N
methoxyacetic acid,acetic acid, methoxy,methoxyethanoic acid,acetic acid, 2-methoxy,ch3och2cooh,methoxyacetlc acid,unii-f11t1h7q7w,methoxy acetic acid,methoxy-acetic acid,ccris 6518
12251
2-methoxyacetic acid
COCC(=O)O
Applications
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Odor Odorless
Flash Point 114°C (237°F)
Quantity 250g
Boiling Point 202°C to 204°C
pH 1.6
Melting Point 8°C
Linear Formula CH3OCH2CO2H
UN Number UN3265
Beilstein 635800
Formula Weight 90.08
Percent Purity 97%
Chemical Name or Material Methoxyacetic acid
Show More Show Less

GHS H Statement
H360-H314-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : AI8650000

TSCA : TSCA