Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetone, 96%

Catalog No. p-3736838
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

Catalog No. AC175730100

Supplier: Thermo Scientific Chemicals 175730100

Only null left
Add to Cart

CAS: 5878-19-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5878-19-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008768
InChI Key CUZLJOLBIRPEFB-UHFFFAOYSA-N
Synonym methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3
PubChem CID 22172
IUPAC Name 1-methoxypropan-2-one
SMILES COCC(C)=O

Specifications

Color Yellow
Density 0.9500g/mL
Boiling Point 118.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.3970 to 1.3990
Linear Formula CH3OCH2COCH3
Quantity 10 g
Beilstein 01, 822
Specific Gravity 0.95
Solubility Information Solubility in water: miscible
Formula Weight 88.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methoxyacetone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-549-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.