Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyamine Hydrochloride, MP Biomedicals™ PROMO

Catalog No. ICN5540505P Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
PubChem CID 521874
IUPAC Name hydrogen O-methylhydroxylamine chloride
SMILES [H+].[Cl-].CON

Specifications

Melting Point 300°F to 304°F
Quantity 5 g
Synonym o-methylhydroxylamine hcl
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride
RTECSNumber NC3980000
Recommended Storage Room Temperature

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.