Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyamine Hydrochloride, MP Biomedicals™ PROMO

Catalog No. ICN5540525P Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN5540525P 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN5540525P Supplier MP Biomedicals, Inc Supplier No. 0215540525FP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
PubChem CID 521874
IUPAC Name hydrogen O-methylhydroxylamine chloride
SMILES [H+].[Cl-].CON

Specifications

Melting Point 300°F to 304°F
Quantity 25 g
Synonym aminooxy methane hydrochloride
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride
RTECSNumber NC3980000
Recommended Storage Room Temperature

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.