Alfa Aesar™ Methyl 3,5-dimethoxybenzoate, 98%

Manufacturer: Alfa Aesar™ A1148609

Quantity 10g

 View more versions of this product

Catalog No. AAA1148609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2150-37-0
    Molecular Formula C10H12O4
    Formula Weight 196.2
    MDL Number MFCD00008432
    Beilstein 2693126
    Synonym 3,5-Dimethoxybenzoic acid methyl ester
    Chemical Name or Material Methyl 3,5-dimethoxybenzoate

    Specifications

    Formula Weight 196.2
    Molecular Formula C10H12O4
    MDL Number MFCD00008432
    Beilstein 2693126
    Synonym 3,5-Dimethoxybenzoic acid methyl ester
    Melting Point 40° to 43°C
    Boiling Point 298°C
    Quantity 10g
    Flash Point >110°C (230°F)
    CAS 2150-37-0
    Chemical Name or Material Methyl 3,5-dimethoxybenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit