Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-bromothiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. p-7055279
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 3-bromothiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26137-08-6
Molecular Formula C6H5BrO2S
Molecular Weight (g/mol) 221.068
MDL Number MFCD00173839
InChI Key PEGSJNCGPSIJOX-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
PubChem CID 2740074
IUPAC Name methyl 3-bromothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)Br

Specifications

Melting Point 50°C to 51°C
Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 221.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-bromothiophene-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.