Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Bromothiophene-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. p-7133309
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 26137-08-6
Molecular Formula C6H5BrO2S
Molecular Weight (g/mol) 221.068
MDL Number MFCD00173839
InChI Key PEGSJNCGPSIJOX-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
PubChem CID 2740074
IUPAC Name methyl 3-bromothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)Br

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 221.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Bromothiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.