Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-hydroxy-4-methoxybenzoate, 98%

Catalog No. ac399260010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399260010 1 g
AC399260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399260010 Supplier Thermo Scientific Chemicals Supplier No. 399260010
Add to Cart

CAS: 6702-50-7 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6702-50-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD01321262
InChI Key QXOXUEFXRSIYSW-UHFFFAOYSA-N
Synonym methyl isovanillate, methyl 3-hydroxy-4-methoxybenzenecarboxylate, 3-hydroxy-4-methoxybenzoic acid methyl ester, isovanillic acid methyl ester, methyl-3-hydroxy-4-methoxybenzoate, methyl 3-hydroxy-4-methoxy-benzoate, benzoic acid, 3-hydroxy-4-methoxy-, methyl ester, methyl 3-hydroxy-4-methoxy benzoate, isovamllic acid methyl ester, ksc496e5l
PubChem CID 4056967
IUPAC Name methyl 3-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC(O)=C(OC)C=C1

Specifications

Melting Point 64.0°C to 67.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)COOCH3
Quantity 1 g
Formula Weight 182.17
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-methoxybenzoate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.