Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxy-3-methoxycinnamate, 99%

Catalog No. AAB2265709
Click to view available options
Quantity:
2 g
10 g

CAS: 2309-07-1 | C16H25ClN2O | 296.84 g/mol

Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2309-07-1
MDL Number MFCD00017208

Specifications

Melting Point 62°C to 65°C
Molecular Formula C11H12O4
Quantity 10 g
Synonym methyl ferulate, methyl 3-4-hydroxy-3-methoxyphenyl acrylate, ferulic acid methyl ester, ferulic acid methylester, methyl 4-hydroxy-3-methoxycinnamate, cinnamic acid, 4-hydroxy-3-methoxy-, methyl ester, unii-y98bua66rx, y98bua66rx, methyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl-, methyl ester
Solubility Information Slightly soluble in water.
InChI Key AUJXJFHANFIVKH-GQCTYLIASA-N
SMILES COC1=C(C=CC(=C1)C=CC(=O)OC)O
IUPAC Name methyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 208.21
PubChem CID 5357283
ChEBI CHEBI:67379
Formula Weight 208.22
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxy-3-methoxycinnamate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber GD9470000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.