Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M285225G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 71486-53-8
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.627
MDL Number MFCD00012769
InChI Key NMAACQILAGCQPR-UHFFFAOYSA-N
Synonym methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
PubChem CID 2724028
IUPAC Name methyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 25 g
Formula Weight 193.63
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.