Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M197525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M197525G 25 g
M19755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M197525G Supplier TCI America Supplier No. M197525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62140-67-4
Molecular Formula C11H14O5S
Molecular Weight (g/mol) 258.288
MDL Number MFCD01317547
InChI Key UUARTHQJSZTOEM-UHFFFAOYSA-N
Synonym 5-(Ethylsulfonyl)-2-methoxybenzoic Acid Methyl Ester
PubChem CID 6454351
IUPAC Name methyl 5-ethylsulfonyl-2-methoxybenzoate
SMILES CCS(=O)(=O)C1=CC(=C(C=C1)OC)C(=O)OC

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 25 g
Formula Weight 258.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.