Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech.

Catalog No. AAA109820E
Click to view available options
Quantity:
500 g
2500 g

CAS: 25134-21-8 | C10H10O3 | 178.19 g/mol

Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

Solubility
Miscible with acetone, benzene, naphtha and xylene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25134-21-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00167036
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym Methyl nadic anhydride

Specifications

Density 1.232
Boiling Point 137°C to 140°C (10 mmHg)
Flash Point 135°C (275°F)
Refractive Index 1.5052
Quantity 2500 g
Sensitivity Moisture sensitive
Solubility Information Miscible with acetone,benzene,naphtha and xylene.
Formula Weight 178.19
Grade Technical
Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers
Hazard Category H302-H315-H317-H318-H331-H334
Hazard Statement GHS H Statement
H301-H334-H332-H315-H319-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P311-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 246-644-8
RTECSNumber RB9100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.