Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, Electronic Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. p-7157734 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
4 x 1 gal.
Packaging:
Poly Bottle

M1239, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, Electronic Grade is also known as wood alcohol and methanol. Spectrum′s ELECTRONIC GRADE products are suitable for use in electronic component manufacturing. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Acidity 0.3 μeq/g
CAS Max % 100%
Alkalinity 0.1 μeq/g
Quantity 4 x 1 gal.
Packaging Poly Bottle
Residue after Evaporation 5 ppm
UN Number UN1230
Water 0.05%
Percent Purity 99.9%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.