Learn More
Methyl-alpha-D-mannopyranoside, 99%
Description
Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.
Solubility
It is easily soluble in methanol, water. Insoluble in ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agent. Stable under recommended storage conditions
Specifications
Specifications
| Melting Point | 193°C to 196°C |
| Quantity | 25 g |
| Beilstein | 81566 |
| Solubility Information | It is easily soluble in methanol,water. Insoluble in ether. |
| Optical Rotation | +79° (c=10 in water) |
| Formula Weight | 194.18 |
| Percent Purity | 99% |
| Chemical Name or Material | Methyl-alpha-D-mannopyranoside |
Safety and Handling
| EINECSNumber | 210-502-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.