Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl benzenesulfonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
Solubility
Miscible with chloroform, methanol, ethanol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
Solubility
Miscible with chloroform, methanol, ethanol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | -4°C |
Density | 1.269 |
Boiling Point | 280°C |
Flash Point | 143°C (289°F) |
Refractive Index | 1.5165 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 908448 |
Solubility Information | Miscible with chloroform,methanol,ethanol and ether. Immiscible with water. |
Formula Weight | 172.2 |
Show More |
Safety and Handling
Hazard Category | H302-H314-H317-H335 |
Hazard Statement | GHS H Statement H314-H341-H302 Causes severe skin burns and eye damage. Suspected of causing genetic defects. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 201-256-8 |
RTECSNumber | DB7151000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction