Learn More
Methyl benzenesulfonate, 98%
Description
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
Solubility
Miscible with chloroform, methanol, ethanol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Specifications
Specifications
Melting Point | -4°C |
Density | 1.269 |
Boiling Point | 280°C |
Flash Point | 143°C (289°F) |
Refractive Index | 1.5165 |
Quantity | 500 g |
UN Number | UN3265 |
Beilstein | 908448 |
Solubility Information | Miscible with chloroform,methanol,ethanol and ether. Immiscible with water. |
Formula Weight | 172.2 |
Show More |
Safety and Handling
Hazard Category | H302-H314-H317-H335 |
Hazard Statement | GHS H Statement H314-H341-H302 Causes severe skin burns and eye damage. Suspected of causing genetic defects. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 201-256-8 |
RTECSNumber | DB7151000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.