Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl benzenesulfonate, 99%

Catalog No. p-160023
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 80-18-2 | C7H8O3S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-18-2
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00014737
InChI Key CZXGXYBOQYQXQD-UHFFFAOYSA-N
Synonym benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
PubChem CID 6630
IUPAC Name methyl benzenesulfonate
SMILES COS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point -4.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 280.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.5160
Quantity 25 g
Beilstein 07,645
Specific Gravity 1.3
Formula Weight 172.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl benzenesulfonate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 201-256-8
RTECSNumber DB7151000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.