Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzoylformate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
15206-55-0
C9H8O3
164.16
MFCD00008443
YLHXLHGIAMFFBU-UHFFFAOYSA-N
methyl benzoylformate,methyl phenylglyoxylate,methyl oxophenylacetate,methyl phenylglyoxalate,benzoylformic acid methyl ester,methylbenzoylformate,phenylglyoxylic acid methyl ester,phenylglyoxylic acid, methyl ester,benzeneacetic acid, .alpha.-oxo-, methyl ester,unii-23f2045stg
84835
CHEBI:84256
methyl 2-oxo-2-phenylacetate
COC(=O)C(=O)C1=CC=CC=C1
15206-55-0
C9H8O3
164.16
MFCD00008443
YLHXLHGIAMFFBU-UHFFFAOYSA-N
methyl benzoylformate,methyl phenylglyoxylate,methyl oxophenylacetate,methyl phenylglyoxalate,benzoylformic acid methyl ester,methylbenzoylformate,phenylglyoxylic acid methyl ester,phenylglyoxylic acid, methyl ester,benzeneacetic acid, .alpha.-oxo-, methyl ester,unii-23f2045stg
84835
CHEBI:84256
methyl 2-oxo-2-phenylacetate
COC(=O)C(=O)C1=CC=CC=C1
Melting Point 16°C
Boiling Point 246°C to 248°C
Flash Point 77°C
Specific Gravity 1.15
Refractive Index 1.5250 to 1.527
Quantity 5g
Density 1.1500g/mL
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5COCO2CH3
Beilstein 10, 657
Solubility Information Solubility in water: 2.02g/L (20°C)
Formula Weight 164.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Methyl benzoylformate, 99%
Show More Show Less

EINECSNumber : 239-263-3

TSCA : TSCA