Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl carbamate, 99%

Catalog No. p-161191
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle

CAS: 598-55-0 | C2H5NO2 | 75.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-55-0
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00007964
InChI Key GTCAXTIRRLKXRU-UHFFFAOYSA-N
Synonym urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech
PubChem CID 11722
ChEBI CHEBI:76606
IUPAC Name methyl carbamate
SMILES COC(N)=O

Specifications

Melting Point 54.0°C to 56.0°C
Boiling Point 176.0°C to 177.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCO2CH3
Beilstein 03, 21
Merck Index 15, 6110
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: very soluble in ethanol,soluble in acetone and ether
Quantity 5 g
Flash Point 93°C
Formula Weight 75.06
Percent Purity 99%
Physical Form Adhering Crystals
Chemical Name or Material Methyl carbamate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 209-939-2
RTECSNumber FC2450000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.