Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Green (Certified Biological Stain), Fisher Chemical

Catalog No. m29525 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M295-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M295-25

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35Cl4N3Zn
Molecular Weight (g/mol) 608.78
MDL Number MFCD00151094
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
Synonym Ethyl Green
PubChem CID 131675247
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]

Specifications

Melting Point 233°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 653.24
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 42585
Color Green
Quantity 25 g
Physical Form Solid
Chemical Name or Material Methyl Green
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.