Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-Lysine Antibody, Novus Biologicals™
SDP

Catalog No. NB600824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600824 Supplier Novus Biologicals Supplier No. NB600824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Methyl-Lysine
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:5000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Alias Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine
Host Species Rabbit
Immunogen Methylated KLH Conjugated detects proteins containing methylated lysine residues in SDS PAGE immunoblots.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects proteins containing methylated lysine residues.
Target Species Non-species specific
Content And Storage Store at -20 °C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.