Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-Lysine Antibody, Novus Biologicals™
SDP

Catalog No. NBP178104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178104 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178104 Supplier Novus Biologicals Supplier No. NBP178104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Methyl-Lysine
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:1000-1:20000, Immunoprecipitation
Formulation 0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0, 50% (v/v) Glycerol with 0.01% Sodium Azide
Gene Alias Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine
Host Species Rabbit
Immunogen This affinity-purified Methyl-Lysine Antibody was prepared from whole rabbit serum produced by repeated immunizations with Methyl-Lysine conjugated KLH.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Test Specificity Reactivity is specific for Methylated Lysine residues (mono, di).
Target Species Non-species specific
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.