Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl-Lysine Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP315955100UL
Description
Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in All species samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifications
Methyl-Lysine | |
Polyclonal | |
Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 | |
Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine | |
A synthetic peptide corresponding to a sequence containing Tri-methylated K. | |
100 μg | |
Phospho Specific | |
All species | |
Purified |
ChIP Assay, Immunoprecipitation | |
Unconjugated | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction