Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl-Lysine Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Methyl-Lysine |
---|---|
Dilution | Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 |
Applications | ChIP Assay, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in All species samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifications
Methyl-Lysine | |
ChIP Assay, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence containing Tri-methylated K. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
All species | |
Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title