Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl methanesulfonate, 99%

Catalog No. AAH5512014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5512014 25 g
AAH5512006 5 g
AAH5512022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5512014 Supplier Thermo Scientific Chemicals Supplier No. H5512014
Only null left
Add to Cart

CAS: 66-27-3 | C2H6O3S | 110.127 g/mol

Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

Solubility
Very soluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-27-3
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
MDL Number MFCD00007557
InChI Key MBABOKRGFJTBAE-UHFFFAOYSA-N
Synonym methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
PubChem CID 4156
ChEBI CHEBI:25255
IUPAC Name methyl methanesulfonate
SMILES COS(=O)(=O)C

Specifications

Color Colorless to Yellow
Density 1.3 g/mL
Boiling Point 202°C to 203°C
Flash Point 104°C (219°F)
Refractive Index 1.414
Linear Formula CH3SO3CH3
Quantity 25 g
UN Number UN2810
Beilstein 1098586
Merck Index 14,6097
Solubility Information Very soluble in water.
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl methanesulfonate
Show More Show Less
Hazard Category H301-H315-H319-H335-H341-H350
Hazard Statement GHS H Statement
H301-H350-H341-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 200-625-0
RTECSNumber PB2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.