Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl methanesulfonate, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.3 g/mL |
| Boiling Point | 202°C to 203°C |
| Flash Point | 104°C (219°F) |
| Refractive Index | 1.414 |
| Linear Formula | CH3SO3CH3 |
| Quantity | 5 g |
| UN Number | UN2810 |
| Beilstein | 1098586 |
| Merck Index | 14,6097 |
| Show More |
Safety and Handling
| Hazard Category | H301-H315-H319-H335-H341-H350 |
| Hazard Statement | GHS H Statement H301-H350-H341-H315-H319-H335 Toxic if swallowed. May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 200-625-0 |
| RTECSNumber | PB2625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction