Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications
Color | Colorless to Yellow |
Density | 1.3 g/mL |
Boiling Point | 202°C to 203°C |
Flash Point | 104°C (219°F) |
Refractive Index | 1.414 |
Linear Formula | CH3SO3CH3 |
Quantity | 5 g |
UN Number | UN2810 |
Beilstein | 1098586 |
Merck Index | 14,6097 |
Show More |
Safety and Handling
Hazard Category | H301-H315-H319-H335-H341-H350 |
Hazard Statement | GHS H Statement H301-H350-H341-H315-H319-H335 Toxic if swallowed. May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
EINECSNumber | 200-625-0 |
RTECSNumber | PB2625000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction