Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-p-benzoquinone, 98%

Catalog No. p-7050385
Click to view available options
Quantity:
10 g
50 g

CAS: 553-97-9 | C7H6O2 | 122.12 g/mol

Methyl-p-benzoquinone imine is the products were the tetrahedral adducts. Synthesis of 2H-indazole-4,7-dione derivatives from3-phenylsydnone and p-toluquinone. A Michael addition of 2-methylcyclopentane-1,3-dione to various p-toluquinone imides exclusively afforded the sole products in a regiospecific manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-97-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00001603
InChI Key VTWDKFNVVLAELH-UHFFFAOYSA-N
Synonym p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone
PubChem CID 11122
IUPAC Name 2-methylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=CC1=O

Specifications

Melting Point 67°C to 70°C
Color Green-Yellow
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Methyl-p-benzoquinone
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 209-056-2
RTECSNumber DK6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.