Learn More
Methyl palmitate, 97%
Description
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.
Solubility
Soluble in ethanol, chloroform, dimethyl sulfoxide and dimethyl formamide. Insoluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 28°C to 34°C |
| Density | 0.852 |
| Boiling Point | 135°C to 137°C (0.2 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.4512 |
| Linear Formula | CH3(CH2)14CO2CH3 |
| Quantity | 100 g |
| Beilstein | 1780973 |
| Solubility Information | Soluble in ethanol,chloroform,dimethyl sulfoxide and dimethyl formamide. Insoluble in water. |
| Formula Weight | 270.46 |
| Show More |
Safety and Handling
| EINECSNumber | 203-966-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.