Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl phenyl sulfone, 98+%

Catalog No. AAA1172709
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAA1172709

Supplier: Thermo Scientific Chemicals A1172709

Only null left
Add to Cart

CAS: 3112-85-4 | C7H8O2S | 156.199 g/mol

Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

Solubility
Soluble in methanol, ethanol, benzene, and chloroform. Partly miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3112-85-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00014741
InChI Key JCDWETOKTFWTHA-UHFFFAOYSA-N
Synonym methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone
PubChem CID 18369
IUPAC Name methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 85°C to 88°C
Quantity 10 g
Beilstein 1906914
Solubility Information Soluble in methanol,ethanol,benzene,and chloroform. Partly miscible in water.
Formula Weight 156.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfone
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 221-476-8
RTECSNumber WR6800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.