Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl phenylsulfonylacetate, 98%

Catalog No. p-7058278
Click to view available options
Quantity:
5 g
25 g

CAS: 34097-60-4 | C9H10O4S | 214.235 g/mol

Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34097-60-4
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00007555
InChI Key NLEAIFBNKPYTGN-UHFFFAOYSA-N
PubChem CID 555619
IUPAC Name methyl 2-(benzenesulfonyl)acetate
SMILES COC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point ∼30°C
Density 1.24
Boiling Point 165°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.535
Quantity 5 g
Beilstein 2051463
Synonym methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate
Solubility Information Insoluble in water.
Formula Weight 214.24
Percent Purity 98%
Chemical Name or Material Methyl phenylsulfonylacetate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.