Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl chloroformate, 98%, AcroSeal™

Catalog No. AC430811000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy

Catalog No. AC430811000

Supplier: Thermo Scientific Chemicals 430811000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 543-27-1 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-27-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000642
InChI Key YOETUEMZNOLGDB-UHFFFAOYSA-N
Synonym isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
PubChem CID 62365
IUPAC Name 2-methylpropyl carbonochloridate
SMILES CC(C)COC(=O)Cl

Specifications

Acidity 0.1% max.
Density 1.0475g/mL
Boiling Point 128.7°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. isobutyl carbonate (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula ClCO2CH2CH(CH3)2
Quantity 100 mL
Merck Index 15,5184
Specific Gravity 1.0475
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes in alcohol,miscible with benzene,chloroform,acetone,toluene
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 208-840-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.