Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), Reagents

Catalog No. p-200021605
Click to view available options
Quantity:
10 g
25 g

Catalog No. 01-336-737

Supplier: Reagents Holdings Llc C611850010C1

Only null left
Add to Cart
Add to Cart

Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O

Specifications

Grade ACS
Packaging Glass Bottle
Specific Gravity 0.79
Color Brown to Red
Quantity 10 g
Physical Form Solid
Chemical Name or Material Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.