Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Solution R, Ricca Chemical
SDP

Catalog No. 504716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5047-16 500 mL Amber Glass Bottle
5047-4 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5047-16 Supplier Ricca Chemical Company Supplier No. 504716
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 67-56-1 , 493-52-7 , 7732-18-5
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) Mixture
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
PubChem CID 10303

Specifications

CAS Min % 55.0, 42.65, 0.05, 0.02
CAS Max % 58.0, 44.39, 0.06, 0.02
Grade Indicator
Packaging Amber Glass Bottle
Color Red
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Methyl Red Solution, Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.