Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Salicylate, FCC, 98%, Spectrum™ Chemical
SDP

Catalog No. 18604452 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle

Catalog No. 18-604-452

Supplier: Spectrum Chemical Manufacturing Corporation M136320LT

Only null left
Add to Cart
Add to Cart

M1363, 119-36-8, C8H8O3

Spectrum™ Chemical Methyl Salicylate, FCC is used as a fragrant in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.535-1.538
Quantity 20 L
Percent Purity 98%
Grade FCC
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.