Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfone, 98%

Catalog No. p-4509823
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

CAS: 67-71-0 | C2H6O2S | 94.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-71-0
Molecular Formula C2H6O2S
Molecular Weight (g/mol) 94.13
MDL Number MFCD00007566
InChI Key HHVIBTZHLRERCL-UHFFFAOYSA-N
Synonym dimethyl sulfone, methyl sulfone, sulfonylbismethane, dimethylsulfone, dimethyl sulphone, methane, sulfonylbis, sulphonylbismethane, methylsulfone, methanesulfonylmethane, sulfonyldimethane
PubChem CID 6213
ChEBI CHEBI:9349
IUPAC Name methylsulfonylmethane
SMILES CS(C)(=O)=O

Specifications

Melting Point 107.0°C to 111.0°C
Color White
Boiling Point 238.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula (CH3)2SO2
Quantity 5 g
Beilstein 01,289
Fieser 01,296; 02,157
Merck Index 15,3283
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in methanol,ethanol and acetone,sparingly soluble in ether
Formula Weight 94.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl sulfone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
EINECSNumber 200-665-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.