Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tert-butyl ether Antibody, Novus Biologicals™
SDP

Catalog No. NB036471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036471 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036471 Supplier Novus Biologicals Supplier No. NBP288852
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Methyl tert-butyl ether Polyclonal specifically detects Methyl tert-butyl ether in Non-species specific samples. It is validated for Immunoaffinity Purification.

Specifications

Antigen Methyl tert-butyl ether
Applications Affinity Purification
Classification Polyclonal
Concentration 5 mg/ml
Conjugate Unconjugated
Formulation 0.1 M PBS (pH 7.2)
Gene Alias METHYL-T-BUTYL ETHER, tert-Butyl methyl ether
Host Species Chicken
Immunogen BSA-tert.Butyl Carbamide
Purification Method PEG purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.