Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tetradecanoate, 99%

Catalog No. AAA1025730
Click to view available options
Quantity:
50 g
250 g

CAS: 124-10-7 | C15H30O2 | 242.40 g/mol

Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.

Solubility
Soluble in chloroform and methanol. React with water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.40
MDL Number MFCD00008983
InChI Key ZAZKJZBWRNNLDS-UHFFFAOYSA-N
Synonym methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
PubChem CID 31284
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 18°C
Density 0.855
Boiling Point 323°C
Flash Point >112°C (234°F)
Refractive Index 1.4562
Linear Formula CH3(CH2)12CO2CH3
Quantity 250 g
Solubility Information Soluble in chloroform and methanol. React with water.
Formula Weight 242.4
Percent Purity 99%
Chemical Name or Material Methyl tetradecanoate
Show More Show Less
EINECSNumber 204-680-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.