Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclohexane, 99%, Reagent Grade, Solstice

Catalog No. 60048083 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-083 20 L Stainless steel can
60-048-081 1 L Glass bottle
60-048-082 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-048-083 Supplier Solstice Supplier No. M3788920L

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Color Colorless
Density 0.77g/mL at 25°C
Boiling Point 100°C to 102°C (1.013 hPa)
Flash Point −4°C (24.8°F)
Odor Characteristic
Packaging Stainless steel can
Linear Formula C6H11CH3
Quantity 20 L
UN Number UN2296
Beilstein 505972
Formula Weight 98.19g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Methylcyclohexane
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
DOTInformation 3/PGII
RTECSNumber 203-624-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.