Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(aminomethyl)benzoate hydrochloride, 97%

Catalog No. AC396780010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC396780010

Supplier: Thermo Scientific Chemicals 396780010

Only null left
Add to Cart

CAS: 6232-11-7 | C9H11NO2·ClH | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6232-11-7
Molecular Formula C9H11NO2·ClH
Molecular Weight (g/mol) 201.65
MDL Number MFCD00182671
InChI Key GIZCKBSSWNIUMZ-UHFFFAOYSA-N
Synonym methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride
PubChem CID 2729253
IUPAC Name methyl 4-(aminomethyl)benzoate;hydrochloride
SMILES COC(=O)C1=CC=C(C=C1)CN.Cl

Specifications

Melting Point 234.5°C to 237.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Linear Formula NH2CH2C6H4COOCH3
Quantity 1 g
Formula Weight 201.65
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(aminomethyl)benzoate hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.