Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylenecyclopentane 98.0+%, TCI America™
SDP

Catalog No. m09141ml Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
M09141ML 1 mL
1 options

Catalog No. M09141ML

Supplier: TCI America M09141ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1528-30-9
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001421
InChI Key NFJPEKRRHIYYES-UHFFFAOYSA-N
PubChem CID 73714
IUPAC Name methylidenecyclopentane
SMILES C=C1CCCC1

Specifications

Color Colorless
Boiling Point 77°C
Quantity 1 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylenecyclopentane
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.