Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog No. p-4229312
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle in Tin Can
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-27-7
Molecular Formula C2H3F3O3S
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000409
InChI Key OIRDBPQYVWXNSJ-UHFFFAOYSA-N
Synonym methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
PubChem CID 9526
IUPAC Name methyl trifluoromethanesulfonate
SMILES COS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.4500g/mL
Boiling Point 95.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.3250 to 1.3270
Linear Formula CF3SO3CH3
Quantity 100 g
Fieser 06,406; 12,329; 13,327; 17,191
Specific Gravity 1.45
Merck Index 15,6204
Formula Weight 164.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-371-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.