Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl vanillate, 99%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Boiling Point | 285.0°C to 287.0°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 5g |
Melting Point | 64.0°C to 67.0°C |
Color | Beige to Yellow |
Assay Percent Range | 98.5% min. (GC) |
Linear Formula | HOC6H3(OCH3)CO2CH3 |
Beilstein | 10, 396 |
Formula Weight | 182.18 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 223-525-9