Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylglyoxal Antibody (9E7), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N259368P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N259368P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N259368P594 Supplier Novus Biologicals Supplier No. NBP259368PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Flow Cytometry.

Specifications

Antigen Methylglyoxal
Applications Flow Cytometry
Classification Monoclonal
Clone 90000000
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 2-oxopropanal, Pyruvaldehyde
Host Species Mouse
Immunogen Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Target Species Human, Non-species specific
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.